Learn More
Encon™ Drench Hose Station
Encon Drench Hose Station provides workers with quick access to flushing during an eye-related emergency.
Manufacturer:  Encon™ 01090103
 View more versions of this product
Catalog No. 18-999-342B
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ANSI Z358.1-2004
CP Bronze stay open ball and CP ball with Stainless Steel valve stem and push plate, Acetal eyewash head, BCP wye and supply pipe mount, ABS Plastic head and Float-off cover secured, coiled hose, wall bracket
1.27cm
45psi
8 gpm
0.5 in.
Pipe
Eyewash hose station