Encon Eyewash Accessories: Bowls

Manufacturer: ENCON SAFETY PRODUCTS 01110462

 View more versions of this product

Catalog No. 19-021778

  Check Availability
Add to Cart

    Specifications

    Product Type Bowl
    Color Yellow
    For Use With (Equipment) Shower and combination eye/facewash
    Material ABS Plastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit