Encon Eyewash Accessories: Float-Off Covers

Manufacturer: ENCON SAFETY PRODUCTS 01052113

 View more versions of this product

Catalog No. 19-021742

  Check Availability
Add to Cart

    Specifications

    Product Type Float-off Cover
    Color Black
    For Use With (Equipment) Shower and combination eye/facewash
    Includes 8 in. Stainless Steel chain
    Material ABS Plastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit