Learn More
Encon™ Fire Blanket Wall Case
Withstands the harshest of conditions, protecting contents so they willl be ready in an emergency
Manufacturer:  Encon™ 01338003
Catalog No. 19-811-395
Add to cart
Description
Description
  • Chemical-resistant ABS plastic thermoformed construction
  • Corrosion-resistant external components
  • Rugged oversize hinges
  • Bulb rubber gasket provides virtual air tight seal
  • Models with stay clear acrylic viewing window(s)
  • Internal dual latch closure with one quarter turn handle
  • High visibility white graphics
  • UV stabilized brilliant red color
  • Suitable for 2.72 to 13.6kg (6 to 30 lb). extinguishers, stored pressure or cartridge operated
  • Reinforced case bottom
  • Fire blanket case compatibility only
Specifications
Specifications
Rugged oversized hinges, bulb Rubber gasket, Internal dual latch closure, reinforced case bottom
Red