Learn More
Encon™ Firststep™ Replacement Bottle for Personal Wash Stations
Manufacturer:  Encon™ 01110787
Catalog No. 19-059473
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For Encon Firstep Dual Eyewash Station (19-035-066). Each bottle contains 32 oz. (946mL) of Firstep, an isotonic, buffered, saline solution with bacteriostatic additive (Hydrosep™) that prevents bacterial growth during prolonged storage.
Specifications
Specifications
946.3mL
Prevents bacterial growth during prolonged storage
32 oz.
Each
Yellow ABS Plastic holder with operational instructions
Firstflush™ Replacement Bottle
For firstep personal eyewash station