Encon™ Modesty Curtain

This water-resistant curtain is yellow for high visibility

Manufacturer: ENCON SAFETY PRODUCTS 01052149

 View more versions of this product

Catalog No. 19-021747

  Check Availability
Add to Cart

    Specifications

    Product Type Modesty Curtain Assembly
    Color Yellow
    For Use With (Equipment) Emergency shower station
    Height (English) 70 in.
    Includes ABS Plastic blocks, Stainless Steel rod, support bracket with Stainless Steel U-bolts
    Material Nylon, PVC
    Height (Metric) 177.8cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit