Learn More
Encon™ Single SCBA Storage Wall Cases
Protects SCBA from potentially damaging and corrosive atmospheres and ensures safe operating capability
Manufacturer:  Encon™ WC60Y00A
 View more versions of this product
Catalog No. 19-811-607
Add to cart
Description
Description
  • Molded plastic construction made of a chemical resistant ABS structural foam
  • Tamper seal security feature-door cannot be opened without breaking the seal
  • Three well spaced interlocking hinges provide excellent support for the door and reduce door deflection when in the open position
  • Bulb rubber gasket provides virtual air tight seal
  • Internal dual latch closure allows for two points of contact to help ensure door seal
  • Quarter turn handle provides for quick and easy access to case contents
  • ABS notched storage shelf for mask/hose attachment
  • Large clear polycarbonate viewing window on lower portion of the door to provide for a SCBA Air Gauge Viewer (AGV)
  • Multiple equipment storage options with variety of shelving options
  • Choice of a fully adjustable steel bracket or Hook-and-Loop strap retaining systems
  • Available with or without high visibility black SCBA graphics
  • Option to select from different colors: Yellow or Green or Safety orange
  • Interior Dimensions: 18.75 x 34.75 x 13.25 in. (47.6 x 88.2 x 33.6cm)
  • Exterior Dimensions: 22 x 37.75 x 13.75 in. (55.8 x 96 x 35cm)
Specifications
Specifications
Molded Plastic
Yellow
Top center/lower shelves
Internal dual latch
Protects SCBA from potentially damaging and corrosive atmospheres and ensures safe operating capability
Single SCBA Storage Wall Case