Learn More
Stay-Open Ball Valve Drench Shower
Encon Stay-Open Ball Valve Drench Shower provides workers with quick access to showering during an emergency.
Manufacturer:  ENCON SAFETY PRODUCTS 01052502
 View more versions of this product
Catalog No. 19-021749
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Vertical shower head
ANSI Z358.1
ABS high visible Plastic shower head, Chrome Plated Bronze stay open ball valve, self adjusting regulators, Galvanized Steel pipe, actuation graphic, sign
1.25 in. NPT
Stainless Steel pull rod
30 gpm
Vertical