Learn More
Encon™ Supply Pipe-Mount Facewash
Self-adjusting regulator which provides 8.0 gpm (30.3Lpm) flow
Manufacturer:  ENCON SAFETY PRODUCTS 01045601
 View more versions of this product
Catalog No. 19-021716
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Facewash
8 gpm
Pipe
ANSI Z358.1
ABS Plastic dual stream head, ABS Plastic Float-off covers, BCP wye, self adjusting regulator, Stainless Steel bead chains, Stainless Steel valve stem, actuation graphic
1.25 in. NPT