Encon™ Wall-Mount Eye Wash

For emergency use, outfit your workspace with the Encon Wall-Mount Eye Wash.

Manufacturer: Encon™ 01110403

 View more versions of this product

Catalog No. 19-021777

  Check Availability
Add to Cart

    Specifications

    Product Type Yellow Bowl
    For Use With (Application) First aid treatment for hazardous material exposure
    Includes ABS Plastic head, wye, push plate, actuation graphic
    Mounting Pipe
    Outlet Connection (English) 1/2 in. NPT
    Pressure Range 30 to 70psi
    Certifications/Compliance ANSI Z358.1-2004
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit