Encon™ Wall-Mount Eye Wash

For emergency use, outfit your workspace with the Encon Wall-Mount Eye Wash.

Manufacturer: Encon™ 01045001

 View more versions of this product

Catalog No. 18-999-339E

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Wall mount fountain
    Flow Rate 2.4 gpm
    Pressure Range 30 to 70psi
    Width (English) 15.62 in.
    Certifications/Compliance ANSI Z358.1-2004
    Color Stainless Steel
    Depth (English) 17.31 in.
    Depth (Metric) 43.96cm
    For Use With (Application) First aid treatment for hazardous material exposure
    Height (English) 9 in.
    Height (Metric) 22.86cm
    Includes Acetal eye/facewash head, BCP wye, Plastic hinge cover, Stainless Steel bowl
    Mounting Wall
    Outlet Connection (English) 1/2 in. NPT
    Width (Metric) 39.67cm
    Activation Stainless Steel push plate

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit