Learn More
Encon™ Wall-Mount Eye Wash
For emergency use, outfit your workspace with the Encon Wall-Mount Eye Wash.
Manufacturer:  Encon™ 01030401
 View more versions of this product
Catalog No. 19-021702
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Eyewash Bottle
ANSI Z358.1-2004
12.62 in.
3.2 gpm
9 in.
High visibility bowl, ABS Plastic head, wye and Float-off covers, Stainless Steel bead chains, self adjusting regulator, Chrome Plated Brass valve
1/2 in. NPT
13.93 in.
Stainless Steel push flag
Yellow
32.05cm
First aid treatment for hazardous material exposure
22.86cm
Wall
30 to 70psi
35.38cm