Learn More
Encon™ Encon™ Yello-Bowl™ Combination Shower/Eye, Facewash without bowl
Made of 80 hot dip galvanized steel piping
Manufacturer:  Encon™ 01050260
Catalog No. 19-811-229
Add to cart
Description
Description
  • Schedule 80 hot dip galvanized steel piping
  • Chrome-plated bronze ball valves and ball
Specifications
Specifications
Galvanized steel
Shower/Eye, Facewash without bowl
20 gpm shower, 8.0 gpm eye/facewash