Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Encon™ Yello-Bowl™ Pedestal-Mount Fountain Eyewashes

Provides first-aid treatment for hazardous material exposure

Manufacturer:  Encon™ 01030305

 View more versions of this product

Catalog No. 19-811-211


Add to cart

Includes: An emergency sign of size 7 x 11 in. (17.8 x 28cm) with legend “EMERGENCY EYEWASH”.

Description

Description

  • ABS plastic heads, wye and float off covers are secured with SS bead chains
  • High visibility ABS plastic, 10.8 in. dia. (27.4cm) Yello-Bowl
  • Self-adjusting regulator to flow at 3.2 gpm (12.1L/min.)
  • Supply connection: 0.5 in. NPT
  • Waste connection: 1.25 in. NPT

  • Model 01030305 comes with a foot treadle

Specifications

Specifications

Pedestal mount fountain
ABS Plastic head, wye, Float-off covers, Stainless Steel bead chains, ABS Plastic yellow-bowl, foot treadle, self adjusting regulator, supply connection, emergency sign
30 to 70psi
3.2 gpm
Pedestal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit