Learn More
Lucigen CorporationSupplier Diversity Partner END-IT DNA END REPAIR KIT DFS Item
Manufacturer:  Lucigen CorporationSupplier Diversity Partner ER81050
End-It Kit converts DNA containing damaged or incompatible 5 and/or 3 -protruding ends to 5-monophosphorylated blunt-ended DNA
Catalog No. NC0105678