Learn More
Sklar InstrumentsSupplier Diversity Partner Kogan Endocervical Speculum, Sklar, Wide Wire Blade, 11 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 903995
Kogan Endocervical Speculum, Sklar, Wide Wire Blade, 11 in., Stainless Steel. Sklar's Kogan Endocervical Speculum is a gynecological instrument used for viewing the distal endocervical canal. Unlike a traditional, duckbill speculum, this single spring, locking, finger-ring intrument features opposing, fenestrated blades at the end of long jaws. The jaws and blades extend deeper into the cervix and can therefore retract deeper tissue. An endocervical speculum is frequently used in colposcopy procedures, or where polyps cannot be easily visualized. This product has a dual screw locking mechanism that adjust along a bar near the finger rings. The bar is gauged with both centimeter and millimeter measurements. This product has wide blades, 4-8mm wide, and an overall length of 11 inches.
Catalog No. 50-143-6714