Learn More
Invitrogen™ ENDOD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552631
Description
Immunogen sequence: SGEDLYILTG TVPSDYRVKD KVAVPEFVWL AACCAVPGGG WAMGFVKHTR DSDIIEDVMV KDLQKLLPFN PQLFQNNCGE TEQDTEKMKK ILEVVNQIQD EERMVQSQKS SSPLSSTRSK RSTLLPPEAS EGSSSFLGKL MGFIATPF Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.
May act as a DNase and a RNase.
Specifications
ENDOD1 | |
Polyclonal | |
Unconjugated | |
ENDOD1 | |
2210414F18Rik; 2310067E08Rik; C85344; Endod1; endonuclease domain containing 1; endonuclease domain-containing 1 protein; Kiaa0830; RGD1310444 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94919 | |
ENDOD1 | |
Recombinant protein corresponding to Human ENDOD1. Recombinant protein control fragment (Product #RP-89926). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.