Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENDOD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549244

Catalog No. PA549244


Add to cart

Description

Description

May act as a DNase and a RNase.
Specifications

Specifications

ENDOD1
Polyclonal
PBS with 0.09% sodium azide
O94919
Endonuclease domain-containing 1 protein, 3130-, ENDOD1, KIAA0830
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23052
Western Blot
Unconjugated
ENDOD1
Liquid
ENDOD1
KLH conjugated synthetic peptide between 255-284 amino acids from the Central region of human ENDOD1.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit