Learn More
Bachem BAM-22P (8-22), 0.5mg, 412961-36-5, MFCD26792549, Trifluoroacetate salt DFS Item
Manufacturer:  Bachem 4047224.0500
Endogenous peptide fragment first isolated from bovine adrenal medulla. BAM-15 is a selective agonist for the sensory neuron specific receptor (EC₅₀ values are 28 and 14 nM for SNSR3 and SNSR4, respectively).
Catalog No. 50-194-6971