Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105 Mouse anti-Human, Alexa Fluor™ 594, Clone: ENG/1326, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254518AF594

Catalog No. NP2418A94


Add to cart

Description

Description

Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human samples. It is validated for Western Blot,Flow Cytometry,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

Endoglin/CD105
ENG/1326
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
ENG
Recombinant human Endoglin protein fragment (aa74-251) (exact sequence is proprietary)
0.1 mL
Primary
2022
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit