Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Endoglin/CD105 Mouse anti-Human, Mouse, Alexa Fluor™ 750, Clone: 3A9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP222122AF750
Catalog No. NP222122A70
Description
Description
Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Human,Mouse samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
Endoglin/CD105
3A9
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
ENG
Purified recombinant fragment of human CD105 expressed in E. coli. [Uniprot: P17813]
0.1 mL
Primary
2022