Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endoglin/CD105, Rabbit anti-Mouse, Clone: MJ7/18, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen Endoglin/CD105
Clone MJ7/18
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Host Species Rabbit
Target Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015691
View Documents
Novus Biologicals
NBP2810850.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015692
View Documents
Novus Biologicals
NBP2810850.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

Endoglin/CD105 Monoclonal antibody specifically detects Endoglin/CD105 in Mouse Samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation.
Specifications

Specifications

Endoglin/CD105
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Mouse
Primary
MJ7/18
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit