Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglycan/PODXL2 Antibody (MM0258- 7R2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212273AF405
Description
Endoglycan/PODXL2 Monoclonal specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot.Specifications
Endoglycan/PODXL2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PODXL2 | |
Human recombinant Endoglycan/PODXL2 (extracellular domain) | |
0.1 mL | |
50512 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
MM0258- 7R2 | |
Western Blot | |
endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction