Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Endoglycan/PODXL2 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0258- 7R2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212273AF350
Catalog No. NP212273A30
Description
Description
Endoglycan/PODXL2 Monoclonal antibody specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Endoglycan/PODXL2
MM0258- 7R2
Western Blot
endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
PODXL2
Human PODXL2
0.1 mL
Primary
50512