Learn More
Endoglycan/PODXL2 Mouse anti-Human, Alexa Fluor 488, Clone: MM0258- 7R2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP212273AF488
Catalog No. NB212273488
Add to cart
Description
Description
Endoglycan/PODXL2 Monoclonal specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
Endoglycan/PODXL2
MM0258- 7R2
Western Blot
Purified
PODXL2
Human PODXL2
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2
Mouse
IgG2
0.1 ml
Primary
50512