Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endoglycan/PODXL2 Mouse anti-Human, Alexa Fluor™ 532, Clone: MM0258- 7R2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212273AF532
Description
Endoglycan/PODXL2 Monoclonal specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Western Blot.Specifications
Endoglycan/PODXL2 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
PODXL2 | |
Human recombinant Endoglycan/PODXL2 (extracellular domain) | |
Primary | |
Human | |
IgG2 |
Western Blot | |
MM0258- 7R2 | |
Western Blot | |
endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2 | |
Mouse | |
0.1 mL | |
50512 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title