Learn More
Alfa Aesar™ Endokinin D, Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66450MA
 View more versions of this product
Catalog No. AAJ66450MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1574.77
Human
Solid
RUO
Endokinin D, human
HPLC
10mg
Val-Gly-Ala-Tyr-Gln-Leu-Glu-His-Thr-Phe-Gln-Gly-Leu-Leu-NH2
-30° to -10°C