Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Endomorphin-2

Click to view available options
Quantity:
5mg
25mg
141801-26-5
C32H37N5O5
571.678
MFCD01321064
XIJHWXXXIMEHKW-LJWNLINESA-N
endomorphin 2,endomorphin-2,tyr-pro-phe-phe-nh2,unii-3ph5m0466g,h-tyr-pro-phe-phe-nh2,l-phenylalaninamide,l-tyrosyl-l-prolyl-l-phenylalanyl,tetrapeptide-15,2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl-n-2s-1-2s-1-amino-1-oxo-3-phenylpropan-2-yl amino-1-oxo-3-phenylpropan-2-yl pyrrolidine-2-carboxamide,tyrosyl-prolyl-phenylalanyl-phenylalaninamide,d00tnr
5311081
(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]-N-[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-1-oxo-3-phenylpropan-2-yl]pyrrolidine-2-carboxamide
C1CC(N(C1)C(=O)C(CC2=CC=C(C=C2)O)N)C(=O)NC(CC3=CC=CC=C3)C(=O)NC(CC4=CC=CC=C4)C(=O)N
141801-26-5
C32H37N5O5
571.678
MFCD01321064
XIJHWXXXIMEHKW-LJWNLINESA-N
endomorphin 2,endomorphin-2,tyr-pro-phe-phe-nh2,unii-3ph5m0466g,h-tyr-pro-phe-phe-nh2,l-phenylalaninamide,l-tyrosyl-l-prolyl-l-phenylalanyl,tetrapeptide-15,2s-1-2s-2-amino-3-4-hydroxyphenyl propanoyl-n-2s-1-2s-1-amino-1-oxo-3-phenylpropan-2-yl amino-1-oxo-3-phenylpropan-2-yl pyrrolidine-2-carboxamide,tyrosyl-prolyl-phenylalanyl-phenylalaninamide,d00tnr
5311081
(2S)-1-[(2S)-2-amino-3-(4-hydroxyphenyl)propanoyl]-N-[(2S)-1-[[(2S)-1-amino-1-oxo-3-phenylpropan-2-yl]amino]-1-oxo-3-phenylpropan-2-yl]pyrrolidine-2-carboxamide
C1CC(N(C1)C(=O)C(CC2=CC=C(C=C2)O)N)C(=O)NC(CC3=CC=CC=C3)C(=O)NC(CC4=CC=CC=C4)C(=O)N
Color White
Quantity 5mg
Formula Weight 571.67
Physical Form Solid
Chemical Name or Material Endomorphin-2

RTECSNumber : SQ7316100

storageNote1 : -30° to -10°C