Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endomucin/Mucin -14 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Endomucin/Mucin -14
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094770
View Documents
Novus Biologicals
NBP303542100UL
100 μg
Each for $320.71
Add to cart
 
NB094771
View Documents
Novus Biologicals
NBP30354220UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Endomucin/Mucin -14 Polyclonal antibody specifically detects Endomucin/Mucin -14 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

Endomucin/Mucin -14
Unconjugated
Rabbit
Human, Mouse
endomucin, MUC-14
Recombinant fusion protein containing a sequence corresponding to amino acids 19-135 of human Endomucin/Mucin -14 (NP_057326.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
51705
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit