Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endophilin A1/SH3GL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | Endophilin A1/SH3GL2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179629
|
Novus Biologicals
NBP179629 |
100 μL |
Each of 1 for $436.00
|
|
Description
Endophilin A1/SH3GL2 Polyclonal specifically detects Endophilin A1/SH3GL2 in Human samples. It is validated for Western Blot.Specifications
Endophilin A1/SH3GL2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CNSA2FLJ25015, EEN-B1bA335L15.1 (SH3-domain GRB2-like 2), Endophilin-1, endophilin-A1, FLJ20276, SH3 domain protein 2A, SH3 domain-containing GRB2-like protein 2, SH3D2AEndophilin A1 BAR domain, SH3-domain GRB2-like 2, SH3P4endophilin-1 | |
SH3GL2 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_003017 | |
6456 | |
Synthetic peptide directed towards the middle region of human SH3GL2The immunogen for this antibody is SH3GL2. Peptide sequence PRREYQPKPRMSLEFPTGDSTQPNGGLSHTGTPKPSGVQMDQPCCRALYD. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title