Learn More
Sklar InstrumentsSupplier Diversity Partner Endoscope Basket, Sklar, 460mm x 225mm x 52mm (18 in. x 9 in. x 2 in.), with 4 Long Silicone Pads (Top/Bottom). Sklar's Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106714
Endoscope Basket, Sklar, 460mm x 225mm x 52mm (18 in. x 9 in. x 2 in.), with 4 Long Silicone Pads (Top/Bottom). Sklar's Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization.
Catalog No. 50-143-7287