Learn More
Sklar InstrumentsSupplier Diversity Partner Endoscope Basket, Sklar, 310mm x 150mm x 77mm (12-1/4 in. x 6 in. x 3 in.), with 3 Silicone Pads (Top/Bottom). Sklar's Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106716
Endoscope Basket, Sklar, 310mm x 150mm x 77mm (12-1/4 in. x 6 in. x 3 in.), with 3 Silicone Pads (Top/Bottom). Sklar's Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization.
Catalog No. 50-143-7288