Learn More
Sklar InstrumentsSupplier Diversity Partner Endoscope Basket, Sklar, 26-1/2 in. x 3-1/4 in. x 2 in. Endoscope Basket 670mm x 80mm x 52mm, with 2 Silicone Pads (Top/Bottom). DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106700
Endoscope Basket, Sklar, 26-1/2 in. x 3-1/4 in. x 2 in. Endoscope Basket 670mm x 80mm x 52mm, with 2 Silicone Pads (Top/Bottom). Sklar's Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization.
Catalog No. 50-143-7280