Learn More
Sklar InstrumentsSupplier Diversity Partner Endoscope Basket, Sklar, 11-1/2 in. x 6-1/4 in. x 2 in. Endoscope Basket #2 ; EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106704
Endoscope Basket, Sklar, 11-1/2 in. x 6-1/4 in. x 2 in. Endoscope Basket #2 290mm x 160mm x 52mm, with 1 Long & 2 Short Silicone Pads (Top/Bottom). Sklar's Endoscope Baskets are designed to store and protect long endoscopic instruments during sterilization.
Catalog No. 50-143-7282