Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Endostatin Antibody (RM0060-3H11), Alexa Fluor™ 488, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212274AF488
Description
Endostatin Monoclonal specifically detects Endostatin in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Endostatin | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COL18A1 | |
Mouse recombinant Endostatin | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
RM0060-3H11 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP | |
Rat | |
Protein A or G purified | |
Angiogenesis, Biologically Active Proteins, Cancer, Extracellular Matrix, Tumor Suppressors | |
80781 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction