Learn More
Endostatin Rat anti-Mouse, Alexa Fluor™ 532, Clone: RM0060-3H11, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212274AF532
Catalog No. NP212274A32
Add to cart
Description
Description
Endostatin Monoclonal antibody specifically detects Endostatin in Mouse samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
Endostatin
RM0060-3H11
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
antiangiogenic agent, collagen alpha-1(XVIII) chain, collagen, type XVIII, alpha 1, FLJ27325, FLJ34914, human type XVIII collagen10endostatin, KNO, KNO1MGC74745, Knobloch syndrome, type 1, KS, multi-functional protein MFP
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
COL18A1
Mouse Endostatin
0.1 mL
Primary
80781