Learn More
Alfa Aesar™ Endothall
A moderately potent inhibitor of protein phosphatase 2A
Manufacturer:  Alfa Aesar™ J60948FPL
 View more versions of this product
Catalog No. AAJ60948FPL
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
20mg
2° to 8°C
A moderately potent inhibitor of protein phosphatase 2A
C8H10O5
7-Oxabicyclo[2.2.1]heptane-2,3-dicarboxylic acid
White
Powder
145-73-3
186.20g/mol
Signal Transduction Reagent