Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Cell Growth Suppl. (Lyoph. Powd. W/ Streptomycin Sulfate), Fisher BioReagents

Specifications

Packaging Amber glass
Storage Requirements 2° to 8°C
Format Powder
Product Type Endothelial Cell Growth Supplement
View More Specs

Products 1
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP265450
View Documents
50mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

Amber glass
2° to 8°C
Powder
Endothelial Cell Growth Supplement
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit