Learn More
Endothelial Cell Growth Supplement, bovine hypothalamus, BT-203, Alfa Aesar™
Our ECGS is used to improve growth, supplement and/or reduce serum concentrations in cultures of endothelial cells, Balb/C-3T3 cells and in hybridoma cell cloning. Optimal concentration range is from 25-200µg/mL.
Manufacturer:  Alfa Aesar J64516MF
 View more versions of this product
Catalog No. AAJ64516MF
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
Lyophilized Powder
50mg
Glass bottle