Learn More
Endothelial Cell Growth Supplement, bovine hypothalamus, BT-203, Alfa Aesar™
Our ECGS is used to improve growth, supplement and/or reduce serum concentrations in cultures of endothelial cells, Balb/C-3T3 cells and in hybridoma cell cloning. Optimal concentration range is from 25-200µg/mL.
Specifications
Format Lyophilized Powder
Packaging Glass bottle
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ64516MF View Documents Alfa Aesar
J64516MF
50mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
AAJ64516AMC View Documents Alfa Aesar
J64516AMC
10 x 50mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications
Specifications
Lyophilized Powder
Glass bottle