Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Endothelial Lipase Mouse anti-Human, PE-Cyanine7, Clone: OTI1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271309PECY7

Catalog No. N271309PEC7


Add to cart

Description

Description

Endothelial Lipase Monoclonal antibody specifically detects Endothelial Lipase in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Endothelial Lipase
OTI1D9
Flow Cytometry
Affinity Purified, Conjugated
LIPG
Full length recombinant protein of human Endothelial Lipase (NP_006024) produced in HEK293T cells.
Immunogen affinity purified
Autophagy, Cholesterol Metabolism, Endothelial Cell Markers, Lipid and Metabolism, Microautophagy, Unfolded Protein Response
Primary
9388.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
EC 3.1.1, EC 3.1.1.3, EDLendothelial lipase, ELlipoprotein lipase H, Endothelial cell-derived lipase, lipase, endothelial, PRO719
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit