Learn More
Alfa Aesar™ Endothelin-1 (1-15)Amide, Human
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66107MA
 View more versions of this product
Catalog No. AAJ66107MA
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1717.04
Human
Solid
RUO
Endothelin-1 (1-15) amide, human
HPLC
10mg
Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-NH2
-30° to -10°C