Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Endothelin A Receptor Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513432
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
This gene encodes the receptor for endothelin-1, a peptide that plays a role in potent and long-lasting vasoconstriction. This receptor associates with guanine-nucleotide-binding (G) proteins, and this coupling activates a phosphatidylinositol-calcium second messenger system. Polymorphisms in this gene have been linked to migraine headache resistance. Alternative splicing results in multiple transcript variants.
Specifications
Endothelin A Receptor | |
Polyclonal | |
Unconjugated | |
EDNRA | |
Ednra; Endor; Endothelin A; endothelin A receptor; endothelin receptor subtype A; endothelin receptor type A; endothelin type A receptor; endothelin-1 receptor; endothelin-1-specific receptor; ETa; ET-A; ETAR; ET-AR; ETA-R; ETRA; G protein coupled receptor; G protein-coupled receptor; Gpcr10; G-protein coupled receptor 10; hET-AR; MFDA; RGD1559432 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
1909 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P25101 | |
EDNRA | |
KLH conjugated synthetic peptide between 125-154 amino acids from the central region of human EDNRA. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction