Learn More
Energy Transformation Apparatus
See how electrical energy is coverted to mechanical energy
Supplier: Science First 6154695
Includes: Assembled motor mounted on plastic base with attached spool and battery clip, length of nylon thread, hook up wires with alligator clips, 15-piece metal washer set, paper clip, and instructions.
Requires : Ammeter, “AA” battery, and a stopwatch.
Description
Use this motor with spool and thread to lift differing numbers of washers to convert one type of energy to another. Electrical input is measured with an ammeter-voltmeter; work is calculated from the mass of the washers and the distance they are raised.
Specifications
Model Thermostat | |
Motor with spool/thread, mounted on Plastic base with attached spool/battery clip, hook up wires with alligator clips, 15-piece metal washer set, paper clip and instructions |