Learn More
Abnova™ ENG 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00002022T01
Catalog No. 89-943-264
Add to cart
Specifications
Specifications
Western Blot
293T
BC014271.2
pCMV-ENG full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
ENG 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 72.49kDa
Transient overexpression cell lysate was tested with anti-ENG antibody (Mfr. No. H00002022-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)