Learn More
ENG Mouse anti-Human, PE, Clone: SN6, Abnova™
Mouse monoclonal antibody raised against ENG.
Manufacturer:  Abnova Corporation MAB12233
Catalog No. 89-934-085
Add to cart
Description
Description
This gene encodes a homodimeric transmembrane protein which is a major glycoprotein of the vascular endothelium. This protein is a component of the transforming growth factor beta receptor complex and it binds TGFB1 and TGFB3 with high affinity. Mutations in this gene cause hereditary hemorrhagic telangiectasia, also known as Osler-Rendu-Weber syndrome 1, an autosomal dominant multisystemic vascular dysplasia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
Specifications
ENG
SN6
Mouse monoclonal antibody raised against ENG.
In PBS, pH7.2 (0.09% Sodium azide, may contain carrier protein/stabilizer).
CD105/END/FLJ41744/HHT1/ORW/ORW1
Mouse
IgG1
100 Reactions
Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE
Flow Cytometry The optimal working dilution should be determined by the end user.
Liquid
ENG
ENG
Affinity Chromatography
RUO
Primary
2022