Learn More
Seracare Life Sciences IncSupplier Diversity Partner ENHANCE BLACK BUFFER SOLUTION  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 55100042
Buffer is mixed with DAB to produce a chromagenic stain that exhibits a black color at the site of reaction. Available as Buffer component of HistoMark Black peroxidase immunohistochemistry substrate kit or as a stand-alone product. Requires DAB solution and Peroxide Solution for use.
Catalog No. 50-675-62