Learn More
Enkurin Mouse anti-Human, Clone: 3B3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524581
Catalog No. PIMA524581
Add to cart
Description
Description
Peptide Sequence: MDPTCSSECI YNLIPSDLKE PPQPPRYISI FKATVKDDMQ KAKTAMKTMG PAKVEVPSPK DFLKKHSKEK TLPPKKNFDR NVPKKPAVPL KTDHPVMGIQ SGKNFINTNA ADIIMGVAKK PKPIYVDKRT GDKHDLEPSG LVPKYINKKD YGVTPEYICK RNEEIKKAQE DYDRYIQENL KKAAMKRLSD EEREAVLQGL KKNWEEVHKE FQSLSVFIDS IPKKIRKQRL EEEMKQLEHD IGIIEKHKII YIANNA
Enkurin interacts with transient receptor potential canonical (TRPC) cation channels (see TRPC1; MIM 602343) and functions as an adaptor protein, tethering signal transduction proteins to TRPC channels (Sutton et al., 2004 [PubMed 15385169]).
Specifications
Specifications
Enkurin
Monoclonal
Unconjugated
ENKUR
Liquid
ENKUR
C10orf63 (NP_659447.1, 1 a.a. ∽ 256 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA, Western Blot
3B3
PBS with no preservative; pH 7.4
Q8TC29
ENKUR; C10orf63
Mouse
IgG2b, kappa
100 μg
-20° C
Primary
219670