Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENL (MLLT1) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$185.00 - $467.00
Specifications
Antigen | ENL (MLLT1) |
---|---|
Concentration | 1.0 mg/mL |
Applications | ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE259625UG
|
MilliporeSigma
ABE259625UG |
25 μg |
Each for $185.00
|
|
|||||
ABE2596100
|
MilliporeSigma
ABE2596100UG |
100 μg |
Each for $467.00
|
|
|||||
Description
ENL (MLLT1) Polyclonal antibody specifically detects ENL (MLLT1) in Human samples. It is validated for ChIP-Seq, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.Specifications
ENL (MLLT1) | |
ChIP sequencing (ChIP-seq), Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
GST-tagged recombinant fragment corresponding to 105 amino acids from the C-terminal half of human ENL (MLLT1) protein. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
1.0 mg/mL | |
Polyclonal | |
Purified | |
RUO | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title