Learn More
Abnova™ ENO1P 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00002025T02
Catalog No. 89-943-273
Add to cart
Specifications
Specifications
Western Blot
293T
BC046928
pCMV-ENO1P full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
ENO1P 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 42.3kDa
Transient overexpression cell lysate was tested with anti-ENO1P antibody (Mfr. No. H00002025-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)